Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.554 in, +0.0002 in, Black Oxide
Catalog No. 5000530071
Encompass
This product is a Class ZZ Plus Plug Gage with a 0.554 inch diameter and a +0.0002 inch tolerance. It features a black oxide finish and is designed for precise measurement applications.
  • Class ZZ Plus Plug Gage
  • Black oxide coating
  • 0.554 inch diameter measurement
  • +0.0002 inch tolerance
Catalog No. 50-005-30071 Supplier MSC Supplier No. 56239056
This product is a Class ZZ Plus Plug Gage with a 0.554 inch diameter and a +0.0002 inch tolerance. It features a black oxide finish and is designed for precise measurement applications.
  • Class ZZ Plus Plug Gage
  • Black oxide coating
  • 0.554 inch diameter measurement
  • +0.0002 inch tolerance