Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5610 in Diameter, +0.0002 in Tolerance, Black Oxide Finish
Catalog No. 5000528797
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.5610 in diameter with a +0.0002 in tolerance and a black oxide finish, making it suitable for checking bore dimensions and maintaining quality control in various industrial settings.
  • Class ZZ Plus Plug Gage
  • 0.5610 in diameter
  • +0.0002 in tolerance
  • Black oxide finish
  • Factory warranty
Catalog No. 50-005-28797 Supplier MSC Supplier No. 56239130
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.5610 in diameter with a +0.0002 in tolerance and a black oxide finish, making it suitable for checking bore dimensions and maintaining quality control in various industrial settings.
  • Class ZZ Plus Plug Gage
  • 0.5610 in diameter
  • +0.0002 in tolerance
  • Black oxide finish
  • Factory warranty