Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.5820 in Dia, +0.000200 in Tolerance, Black Oxide
Catalog No. 5000527776
Encompass
This SPI Class ZZ Plus Plug Gage is designed for precision Go Inspection and for calibrating measuring devices such as micrometers and verniers. It features a 0.5820 in diameter with a tolerance of +0.000200 in (0.0002 mm). The gage has a black oxide finish and includes a Certificate of Accuracy with NIST Traceability, adhering to ASME B89.1 standards.
  • 0.5820 in diameter
  • +0.000200 in (0.0002 mm) tolerance
  • Black oxide finish
  • Suitable for go inspection
  • Used for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards
Catalog No. 50-005-27776 Supplier MSC Supplier No. 56239346
This SPI Class ZZ Plus Plug Gage is designed for precision Go Inspection and for calibrating measuring devices such as micrometers and verniers. It features a 0.5820 in diameter with a tolerance of +0.000200 in (0.0002 mm). The gage has a black oxide finish and includes a Certificate of Accuracy with NIST Traceability, adhering to ASME B89.1 standards.
  • 0.5820 in diameter
  • +0.000200 in (0.0002 mm) tolerance
  • Black oxide finish
  • Suitable for go inspection
  • Used for setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Adheres to ASME B89.1 standards