Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.596 in, +0.0002 in, Black Oxide
Catalog No. 5000531280
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications. It features a 0.596 inch diameter and a +0.0002 inch tolerance, with a durable black oxide finish.
  • Black oxide finish.
  • +0.0002 inch tolerance.
  • 0.596 inch diameter.
Catalog No. 50-005-31280 Supplier MSC Supplier No. 56239486
This Class ZZ Plus plug gage is designed for precise measurement applications. It features a 0.596 inch diameter and a +0.0002 inch tolerance, with a durable black oxide finish.
  • Black oxide finish.
  • +0.0002 inch tolerance.
  • 0.596 inch diameter.