Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.6080 in Diameter, +0.000200 in Tolerance, Black Oxide
Catalog No. 5000527724
Encompass
This Class ZZ Plus Plug Gage features a 0.6080 in diameter and a +0.000200 in tolerance, with a durable black oxide finish. It is designed for precise go inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection
  • Ideal for setting micrometers
  • Can be used for setting verniers and other measuring devices
Catalog No. 50-005-27724 Supplier MSC Supplier No. 56239601
Add to Cart
Add to Cart
This Class ZZ Plus Plug Gage features a 0.6080 in diameter and a +0.000200 in tolerance, with a durable black oxide finish. It is designed for precise go inspection and for setting micrometers, verniers, and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for go inspection
  • Ideal for setting micrometers
  • Can be used for setting verniers and other measuring devices