Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.638 inch Diameter, Black Oxide
Catalog No. 5000527661
Encompass
This Class ZZ Plus Plug Gage features a 0.638 inch diameter and a black oxide finish, designed for precise measurement applications.
  • Class ZZ Plus plug gage
  • Black oxide finish
  • Tolerance of +0.000200 inch
  • Diameter of 0.638 inch
Catalog No. 50-005-27661 Supplier MSC Supplier No. 56239908
This Class ZZ Plus Plug Gage features a 0.638 inch diameter and a black oxide finish, designed for precise measurement applications.
  • Class ZZ Plus plug gage
  • Black oxide finish
  • Tolerance of +0.000200 inch
  • Diameter of 0.638 inch