Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.6390 in, +0.000200 in, Black Oxide
Catalog No. 5000527495
Encompass
This product is a Class ZZ Plus Plug Gage designed for precise measurement applications. It features a 0.6390 in diameter and a tolerance of +0.000200 in, with a durable black oxide finish.
  • Hassle-free guarantee for exchanges or refunds within 30 days of shipment in original packaging.
  • Products are of first quality.
Catalog No. 50-005-27495 Supplier MSC Supplier No. 56239916
This product is a Class ZZ Plus Plug Gage designed for precise measurement applications. It features a 0.6390 in diameter and a tolerance of +0.000200 in, with a durable black oxide finish.
  • Hassle-free guarantee for exchanges or refunds within 30 days of shipment in original packaging.
  • Products are of first quality.