Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.6560 in, +0.000200 in, Black Oxide
Catalog No. 5000527464
Encompass
The product is a Class ZZ Plus Plug Gage with a diameter of 0.6560 in and a tolerance of +0.000200 in. It features a black oxide coating/finish.
  • Class ZZ Plus Plug Gage design
  • Diameter of measurement: 0.6560 in
  • Tolerance of +0.000200 in
  • Black oxide coating or finish
  • Manufactured in China
Catalog No. 50-005-27464 Supplier MSC Supplier No. 56240088
The product is a Class ZZ Plus Plug Gage with a diameter of 0.6560 in and a tolerance of +0.000200 in. It features a black oxide coating/finish.
  • Class ZZ Plus Plug Gage design
  • Diameter of measurement: 0.6560 in
  • Tolerance of +0.000200 in
  • Black oxide coating or finish
  • Manufactured in China