Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7120 in Diameter, 0.0002 in Tolerance, Black Oxide
Catalog No. 5000528802
Encompass
This plug gage is engineered for precise measurement and inspection applications. It features a specific diameter and a tight tolerance, along with a protective black oxide finish for durability.
  • 0.7120 in diameter
  • +0.0002 in tolerance
  • Black oxide finish
Catalog No. 50-005-28802 Supplier MSC Supplier No. 56240658
This plug gage is engineered for precise measurement and inspection applications. It features a specific diameter and a tight tolerance, along with a protective black oxide finish for durability.
  • 0.7120 in diameter
  • +0.0002 in tolerance
  • Black oxide finish