Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7200 in Diameter, 0.000200 in Tolerance
Catalog No. 5000530835
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. Manufactured from durable steel, it ensures accuracy with a specific diameter and tolerance, making it suitable for quality control and calibration tasks.
  • NIST traceable
  • Constructed from steel
  • Features a 0.7200 inch diameter
  • Offers a tight tolerance of 0.000200 inch
Catalog No. 50-005-30835 Supplier MSC Supplier No. 78389053
This Class ZZ Plus Plug Gage is designed for precise measurement applications. Manufactured from durable steel, it ensures accuracy with a specific diameter and tolerance, making it suitable for quality control and calibration tasks.
  • NIST traceable
  • Constructed from steel
  • Features a 0.7200 inch diameter
  • Offers a tight tolerance of 0.000200 inch