Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7220 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000531062
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications. It features a 0.7220 inch diameter and a 0.000200 inch tolerance, making it suitable for checking hole sizes and other critical dimensions.
  • Made from steel for durability.
  • Features a 0.7220 inch diameter for accurate measurements.
  • Provides a precise 0.000200 inch tolerance.
  • Includes a factory warranty.
Catalog No. 50-005-31062 Supplier MSC Supplier No. 78389079
This Class ZZ Plus plug gage is designed for precise measurement applications. It features a 0.7220 inch diameter and a 0.000200 inch tolerance, making it suitable for checking hole sizes and other critical dimensions.
  • Made from steel for durability.
  • Features a 0.7220 inch diameter for accurate measurements.
  • Provides a precise 0.000200 inch tolerance.
  • Includes a factory warranty.