Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7300 in Dia, +0.000200 in Tolerance, Black Oxide
Catalog No. 5000530680
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.7300 in diameter with a tight tolerance of +0.000200 in and a durable black oxide finish.
  • Diameter of measurement: 0.7300 in
  • Tolerance: +0.000200 in
  • Coating/finish: black oxide
  • Country of origin: China
Catalog No. 50-005-30680 Supplier MSC Supplier No. 56240849
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.7300 in diameter with a tight tolerance of +0.000200 in and a durable black oxide finish.
  • Diameter of measurement: 0.7300 in
  • Tolerance: +0.000200 in
  • Coating/finish: black oxide
  • Country of origin: China