Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7510 in Dia, +0.000200 in Tolerance, Black Oxide
Catalog No. 5000530079
Encompass
A Class ZZ Plus Plug Gage is designed for precise measurement applications. This gage features a 0.7510 inch diameter and a tolerance of +0.000200 inch, finished with black oxide for durability.
  • Class ZZ Plus plug gage
  • 0.7510 inch diameter
  • +0.000200 inch tolerance
  • Black oxide finish
Catalog No. 50-005-30079 Supplier MSC Supplier No. 56241052
A Class ZZ Plus Plug Gage is designed for precise measurement applications. This gage features a 0.7510 inch diameter and a tolerance of +0.000200 inch, finished with black oxide for durability.
  • Class ZZ Plus plug gage
  • 0.7510 inch diameter
  • +0.000200 inch tolerance
  • Black oxide finish