Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.7990 in, 0.000200 in Tolerance, Steel
Catalog No. 5000530833
Encompass
This Class ZZ Plus Plug Gage features a 0.7990 inch diameter and a 0.000200 inch tolerance. Manufactured by SPI, this gage is constructed from durable steel.
  • Accurately measures with a 0.7990 inch diameter
  • Features a precise 0.000200 inch tolerance
  • Constructed from durable steel for longevity
Catalog No. 50-005-30833 Supplier MSC Supplier No. 78389848
This Class ZZ Plus Plug Gage features a 0.7990 inch diameter and a 0.000200 inch tolerance. Manufactured by SPI, this gage is constructed from durable steel.
  • Accurately measures with a 0.7990 inch diameter
  • Features a precise 0.000200 inch tolerance
  • Constructed from durable steel for longevity