Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.8100 in, +0.000200 in Tolerance, Black Oxide
Catalog No. 5000530219
Encompass
This Class ZZ Plus Plug Gage is designed for precise dimensional inspection. It features a 0.8100 inch diameter with a tight +0.000200 inch tolerance and a durable black oxide finish.
  • Precise dimensional inspection
  • 0.8100 inch diameter
  • +0.000200 inch tolerance
  • Durable black oxide finish
Catalog No. 50-005-30219 Supplier MSC Supplier No. 56241656
This Class ZZ Plus Plug Gage is designed for precise dimensional inspection. It features a 0.8100 inch diameter with a tight +0.000200 inch tolerance and a durable black oxide finish.
  • Precise dimensional inspection
  • 0.8100 inch diameter
  • +0.000200 inch tolerance
  • Durable black oxide finish