Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.844, +0.0002, Black Oxide
Catalog No. 5000530441
Encompass
This Class ZZ Plus Plug Gage has a 0.844 inch diameter and a +0.0002 inch tolerance. It features a black oxide finish.
  • Black oxide finish
  • +0.0002 inch tolerance
  • 0.844 inch diameter
Catalog No. 50-005-30441 Supplier MSC Supplier No. 56241995
This Class ZZ Plus Plug Gage has a 0.844 inch diameter and a +0.0002 inch tolerance. It features a black oxide finish.
  • Black oxide finish
  • +0.0002 inch tolerance
  • 0.844 inch diameter