Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.9050 in, +0.000200 in Tolerance
Catalog No. 5000530215
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.9050 inch diameter and a +0.000200 inch tolerance, with a durable black oxide coating for enhanced performance.
  • Diameter of 0.9050 "
  • Tolerance of +0.000200 "
  • Features a black oxide coating.
  • Manufactured in China.
Catalog No. 50-005-30215 Supplier MSC Supplier No. 56242605
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.9050 inch diameter and a +0.000200 inch tolerance, with a durable black oxide coating for enhanced performance.
  • Diameter of 0.9050 "
  • Tolerance of +0.000200 "
  • Features a black oxide coating.
  • Manufactured in China.