Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.9110 in Diameter, Black Oxide Finish
Catalog No. 5000529046
Encompass
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.9110 inch diameter with a tight tolerance of +0.000200 inch, ensuring accuracy for critical inspections. The black oxide finish provides enhanced durability and protection.
  • Precise 0.9110 inch diameter for accurate measurements.
  • High tolerance of +0.000200 inch for critical applications.
  • Black oxide finish for improved durability and corrosion resistance.
  • Manufactured to Class ZZ standards.
Catalog No. 50-005-29046 Supplier MSC Supplier No. 56242662
This Class ZZ Plus Plug Gage is designed for precise measurement applications. It features a 0.9110 inch diameter with a tight tolerance of +0.000200 inch, ensuring accuracy for critical inspections. The black oxide finish provides enhanced durability and protection.
  • Precise 0.9110 inch diameter for accurate measurements.
  • High tolerance of +0.000200 inch for critical applications.
  • Black oxide finish for improved durability and corrosion resistance.
  • Manufactured to Class ZZ standards.