Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.9180 in Diameter, Tool Steel, ASME B89.1
Catalog No. 5000511983
Encompass
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection tasks, ensuring accuracy when setting micrometers, verniers, and other measuring devices. Featuring a 0.9180 inch diameter and a tight +0.000200 inch tolerance, it provides reliable performance for quality control applications.
  • Diameter of measurement: 0.9180 inch
  • Tolerance: +0.000200 inch
  • Black oxide coating/finish
  • Constructed from tool steel
  • Suitable for go inspection
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Country of origin: China
Catalog No. 50-005-11983 Supplier MSC Supplier No. 56242746
This Class ZZ Plus Plug Gage is engineered for precise measurement and inspection tasks, ensuring accuracy when setting micrometers, verniers, and other measuring devices. Featuring a 0.9180 inch diameter and a tight +0.000200 inch tolerance, it provides reliable performance for quality control applications.
  • Diameter of measurement: 0.9180 inch
  • Tolerance: +0.000200 inch
  • Black oxide coating/finish
  • Constructed from tool steel
  • Suitable for go inspection
  • Includes certificate of accuracy with NIST traceability
  • Complies with ASME B89.1 standards
  • Country of origin: China