Learn More
MSC SPI Class ZZ Plus Plug Gage, 0.9300 in Diameter, 0.000200 in Tolerance, Steel
Catalog No. 5000528701
Encompass
This plug gage is designed for precise measurement applications. It features a specific diameter and tolerance, manufactured from durable steel to ensure accuracy and longevity in various industrial settings.
  • Diameter of measurement: 0.9300 in.
  • Precise tolerance of 0.000200 in.
  • Constructed from steel for durability.
Catalog No. 50-005-28701 Supplier MSC Supplier No. 78391158
This plug gage is designed for precise measurement applications. It features a specific diameter and tolerance, manufactured from durable steel to ensure accuracy and longevity in various industrial settings.
  • Diameter of measurement: 0.9300 in.
  • Precise tolerance of 0.000200 in.
  • Constructed from steel for durability.