Learn More
MSC SPI CMM Ball Stylus, 2 mm Ball Diameter, 20 mm Overall Length, M2 Thread, Ruby
Catalog No. 5000529703
Encompass
The stylus is the spherical portion or tip of the probe that makes contact with the workpiece surface. It is commonly made of a synthetic ruby and mounted on a stem.
  • Makes contact with the workpiece surface
  • Spherical portion or tip of the probe
  • Commonly made of synthetic ruby
  • Mounted on a stem
  • Ball diameter 2 mm
  • Overall length 20 mm
  • M2 thread size
  • Ruby point material
Catalog No. 50-005-29703 Supplier MSC Supplier No. 06424576
The stylus is the spherical portion or tip of the probe that makes contact with the workpiece surface. It is commonly made of a synthetic ruby and mounted on a stem.
  • Makes contact with the workpiece surface
  • Spherical portion or tip of the probe
  • Commonly made of synthetic ruby
  • Mounted on a stem
  • Ball diameter 2 mm
  • Overall length 20 mm
  • M2 thread size
  • Ruby point material