Learn More
MSC SPI CMM Ball Stylus, 4 mm Ball, 20 mm Overall Length, M2 Thread, Ruby
Catalog No. 5000514040
Encompass
The stylus is the spherical portion or tip of the probe that makes contact with the workpiece surface. It is commonly made of a synthetic ruby and mounted on a stem.
  • Spherical tip design
  • Makes contact with workpiece surface
  • Made of synthetic ruby
  • Mounted on a stem
Catalog No. 50-005-14040 Supplier MSC Supplier No. 06424568
The stylus is the spherical portion or tip of the probe that makes contact with the workpiece surface. It is commonly made of a synthetic ruby and mounted on a stem.
  • Spherical tip design
  • Makes contact with workpiece surface
  • Made of synthetic ruby
  • Mounted on a stem