Learn More
MSC SPI CMM Disc Stylus, 3 mm Ball Diameter, 13 mm Overall Length, M2 Thread
Catalog No. 5000529889
Encompass
This CMM Disc Stylus is designed for making contact with workpiece surfaces. Featuring a 3 mm ball diameter and an overall length of 13 mm, it connects via M2 threads and utilizes a durable steel point for precise measurements.
  • Designed for making contact with workpiece surfaces.
  • Features a 3 mm ball diameter.
  • Disc stylus type.
  • M2 thread size for connection.
  • Steel point material for durability.
  • 13 mm overall length.
Catalog No. 50-005-29889 Supplier MSC Supplier No. 06424642
This CMM Disc Stylus is designed for making contact with workpiece surfaces. Featuring a 3 mm ball diameter and an overall length of 13 mm, it connects via M2 threads and utilizes a durable steel point for precise measurements.
  • Designed for making contact with workpiece surfaces.
  • Features a 3 mm ball diameter.
  • Disc stylus type.
  • M2 thread size for connection.
  • Steel point material for durability.
  • 13 mm overall length.