Learn More
MSC SPI CNC Deluxe Tool Setting Gage, 360 Degree Adjustable, For CNC Slant Bed Machines
Catalog No. 5000531620
Encompass
This CNC Deluxe Tool Setting Gage is designed for use on CNC slant bed machines. It accurately sets turning, boring, and facing tools on the centerline of the workpiece.
  • Designed for use on CNC slant bed machines.
  • Accurately sets turning, boring and facing tools on the centerline of the workpiece.
  • Adjustable to any slant angle (360°).
  • Includes easy-to-read laser etched protractor.
Catalog No. 50-005-31620 Supplier MSC Supplier No. 76841923
This CNC Deluxe Tool Setting Gage is designed for use on CNC slant bed machines. It accurately sets turning, boring, and facing tools on the centerline of the workpiece.
  • Designed for use on CNC slant bed machines.
  • Accurately sets turning, boring and facing tools on the centerline of the workpiece.
  • Adjustable to any slant angle (360°).
  • Includes easy-to-read laser etched protractor.