Learn More
MSC SPI Combination Square Blade, 24 in Length, 4R Graduation, Steel
Catalog No. 5000534921
Encompass
This combination square blade is designed for precise measuring and inspecting tasks. It features a baked enamel coated finish for enhanced durability. The blade includes clear graduations in 1/64, 1/32, 1/16, and 1/8 inch increments. Constructed from steel, this blade is 1 inch wide and 0.09 inches thick, making it a robust tool for various applications.
  • 24 inch length
  • 4R graduation style
  • Baked enamel coated finish
  • Graduations in 1/64, 1/32, 1/16, and 1/8 inch increments
  • Constructed from steel
  • 1 inch blade width
  • 0.09 inch blade thickness
Catalog No. 50-005-34921 Supplier MSC Supplier No. 60591930
This combination square blade is designed for precise measuring and inspecting tasks. It features a baked enamel coated finish for enhanced durability. The blade includes clear graduations in 1/64, 1/32, 1/16, and 1/8 inch increments. Constructed from steel, this blade is 1 inch wide and 0.09 inches thick, making it a robust tool for various applications.
  • 24 inch length
  • 4R graduation style
  • Baked enamel coated finish
  • Graduations in 1/64, 1/32, 1/16, and 1/8 inch increments
  • Constructed from steel
  • 1 inch blade width
  • 0.09 inch blade thickness