Learn More
MSC SPI Combination Square Set, 4 Piece, 18 in Blade, 4R Graduation
Catalog No. 5000527289
Encompass
The SPI 4 Piece, 18 in Combination Square Set is designed for precise measurement applications. It features a hardened steel blade and cast iron center, protractor, and square heads.
  • Comes as a 4-piece set
  • Features an 18 in blade length
  • Equipped with 4R graduation style
  • Includes a hardened steel blade for durability
  • Heads are made from cast iron for robust performance
Catalog No. 50-005-27289 Supplier MSC Supplier No. 60591567
Add to Cart
Add to Cart
The SPI 4 Piece, 18 in Combination Square Set is designed for precise measurement applications. It features a hardened steel blade and cast iron center, protractor, and square heads.
  • Comes as a 4-piece set
  • Features an 18 in blade length
  • Equipped with 4R graduation style
  • Includes a hardened steel blade for durability
  • Heads are made from cast iron for robust performance