Learn More
MSC SPI Truncated Taperlock Thread Setting Plug Gage, #10-24 UNC, Tool Steel, 1 ea
Catalog No. 5000510380
Encompass
This truncated taperlock thread gage set is designed for setting adjustable thread ring gauges. Manufactured from high-quality tool steel, the set provides reliable and precise performance for inspection applications compliant with industry standards.
  • Includes go and no go plug members
  • Includes handle
  • Constructed from durable tool steel
  • Features X tolerance on pitch diameter
  • Features W tolerance on flank angles and lead
  • Certified with inspection certificate traceable to NIST
  • Meets ANSI/ASME B1.2 standards
  • Thread size: #10-24 UNC, class 2A
Catalog No. 50-005-10380 Supplier MSC Supplier No. 46553277
Add to Cart
Add to Cart
This truncated taperlock thread gage set is designed for setting adjustable thread ring gauges. Manufactured from high-quality tool steel, the set provides reliable and precise performance for inspection applications compliant with industry standards.
  • Includes go and no go plug members
  • Includes handle
  • Constructed from durable tool steel
  • Features X tolerance on pitch diameter
  • Features W tolerance on flank angles and lead
  • Certified with inspection certificate traceable to NIST
  • Meets ANSI/ASME B1.2 standards
  • Thread size: #10-24 UNC, class 2A