Learn More
MSC SPI Depth and Angle Gage, 0 to 6 inch Rule, 30 to 60 degree Angle, Hardened Steel Head
Catalog No. 5000527448
Encompass
This combination depth and angle gage provides precise measurements for various applications. It offers a rule measurement range from 0 to 6 inches and an angle measurement range of 30 to 60 degrees. The head is made from hardened steel and the rule from stainless steel, ensuring durability and long-lasting performance.
  • Hardened steel head with 2-1/2 inch base.
  • Stainless steel blade with 32nds and 64ths graduations.
  • 60 degree angle graduation for precise angle measurements.
  • Measures angles at 30, 45, and 60 degrees on both sides.
  • Includes a non-graduated rod for measuring holes under 3/32 inch diameter.
Catalog No. 50-005-27448 Supplier MSC Supplier No. 67698548
This combination depth and angle gage provides precise measurements for various applications. It offers a rule measurement range from 0 to 6 inches and an angle measurement range of 30 to 60 degrees. The head is made from hardened steel and the rule from stainless steel, ensuring durability and long-lasting performance.
  • Hardened steel head with 2-1/2 inch base.
  • Stainless steel blade with 32nds and 64ths graduations.
  • 60 degree angle graduation for precise angle measurements.
  • Measures angles at 30, 45, and 60 degrees on both sides.
  • Includes a non-graduated rod for measuring holes under 3/32 inch diameter.