Learn More
MSC SPI Dial Bore Gage, 0.4 to 0.7 in Diameter, 4 in Probe Depth
Catalog No. 5000527942
Encompass
This Dial Bore Gage is designed for close tolerance measurements of holes, taper, and roundness. It features an interchangeable anvil for accurate dimensional setting and offers a wide measuring range with a sub-anvil. A short form Calibration Certificate is included, and the product comes furnished in a fitted case.
  • Designed for close tolerance measurements of holes, taper, and roundness.
  • Features an interchangeable anvil for accurate dimensional setting.
  • Offers a wide measuring range with a sub-anvil.
  • Includes a short form Calibration Certificate.
  • Furnished in a fitted case.
  • Supplied with anvils, spacers, sub-anvils, and an AGD 2 dial indicator.
  • Two year limited warranty.
Catalog No. 50-005-27942 Supplier MSC Supplier No. 37997541
This Dial Bore Gage is designed for close tolerance measurements of holes, taper, and roundness. It features an interchangeable anvil for accurate dimensional setting and offers a wide measuring range with a sub-anvil. A short form Calibration Certificate is included, and the product comes furnished in a fitted case.
  • Designed for close tolerance measurements of holes, taper, and roundness.
  • Features an interchangeable anvil for accurate dimensional setting.
  • Offers a wide measuring range with a sub-anvil.
  • Includes a short form Calibration Certificate.
  • Furnished in a fitted case.
  • Supplied with anvils, spacers, sub-anvils, and an AGD 2 dial indicator.
  • Two year limited warranty.