Learn More
MSC SPI Dial Bore Gage, 0.7 to 1.5 in Dia, 6 in Probe Depth, 0.000600 in Accuracy
Catalog No. 5000532401
Encompass
This dial bore gage is designed for close tolerance measurements of holes, taper, and roundness. It features an interchangeable anvil attached to a mic head for accurate dimensional setting and offers a wide measuring range with a sub-anvil.
  • For close tolerance measurements of holes, taper, and roundness
  • Interchangeable anvil is attached to a mic head for accurate dimensional setting
  • Wide measuring range with sub-anvils
  • Short form calibration certificate included
  • Furnished in a fitted case
  • Two year limited warranty
Catalog No. 50-005-32401 Supplier MSC Supplier No. 37997715
This dial bore gage is designed for close tolerance measurements of holes, taper, and roundness. It features an interchangeable anvil attached to a mic head for accurate dimensional setting and offers a wide measuring range with a sub-anvil.
  • For close tolerance measurements of holes, taper, and roundness
  • Interchangeable anvil is attached to a mic head for accurate dimensional setting
  • Wide measuring range with sub-anvils
  • Short form calibration certificate included
  • Furnished in a fitted case
  • Two year limited warranty