Learn More
MSC SPI Dial Bore Gage, 0.7 to 1.5 inch, 6 inch Probe, 10 Anvils
Catalog No. 5000512411
Encompass
This dial bore gage is designed for close tolerance measurements of holes, taper, and roundness. It features an interchangeable anvil attached to a mic head for accurate dimensional setting, and offers a wide measuring range with a sub-anvil. The gage is furnished in a fitted case and includes a short form calibration certificate.
  • For close tolerance measurements of holes, taper, and roundness.
  • Interchangeable anvil is attached to a mic head for accurate dimensional setting.
  • Wide measuring range with sub-anvil.
  • Short form calibration certificate included.
  • Furnished in fitted case.
  • Two year limited warranty.
  • Supplied with anvils, spacers, sub-anvils, and an AGD 2 dial indicator.
Catalog No. 50-005-12411 Supplier MSC Supplier No. 37997533
This dial bore gage is designed for close tolerance measurements of holes, taper, and roundness. It features an interchangeable anvil attached to a mic head for accurate dimensional setting, and offers a wide measuring range with a sub-anvil. The gage is furnished in a fitted case and includes a short form calibration certificate.
  • For close tolerance measurements of holes, taper, and roundness.
  • Interchangeable anvil is attached to a mic head for accurate dimensional setting.
  • Wide measuring range with sub-anvil.
  • Short form calibration certificate included.
  • Furnished in fitted case.
  • Two year limited warranty.
  • Supplied with anvils, spacers, sub-anvils, and an AGD 2 dial indicator.