Learn More
MSC SPI Dial Caliper, 6 in, 0.001 in Graduation, 1 ea
Catalog No. 501920069
Encompass
This dial caliper features stainless steel construction and is designed for precise 4-way measurements, including inside, outside, depth, and step. It offers accurate readings with a clear dial and convenient zero adjustment.
  • Stainless steel construction with all measuring surfaces, surface-hardened
  • Preloaded dual pinions eliminate backlash and provide better repeatability
  • Four-way measurements: inside, outside, depth and step
  • Easy zero adjustment per revolution
  • Adjustable bezel with locking device
  • Covered rack keeps gear teeth clean
  • Thumb roller included
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty
Catalog No. 50-192-0069 Supplier MSC Supplier No. 37530748
This dial caliper features stainless steel construction and is designed for precise 4-way measurements, including inside, outside, depth, and step. It offers accurate readings with a clear dial and convenient zero adjustment.
  • Stainless steel construction with all measuring surfaces, surface-hardened
  • Preloaded dual pinions eliminate backlash and provide better repeatability
  • Four-way measurements: inside, outside, depth and step
  • Easy zero adjustment per revolution
  • Adjustable bezel with locking device
  • Covered rack keeps gear teeth clean
  • Thumb roller included
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty