Learn More
MSC SPI Dial Caliper, 18 in, 0.001 in Graduation, Stainless Steel Jaws
Catalog No. 5000528160
Encompass
The dial caliper is a precision measuring instrument designed for accurate inside, outside, depth, and step measurements. Constructed from stainless steel, it offers an 18-inch maximum measurement range with a 0.001 inch graduation and 0.002 inch accuracy. This caliper features preloaded dual pinions for consistent repeatability and comes with NIST traceability certification.
  • Stainless steel construction with surface-hardened measuring surfaces
  • Preloaded dual pinions eliminate backlash and provide better repeatability
  • 4-Way measurements: inside, outside, depth and step
  • Easy zero adjustment per revolution
  • Adjustable bezel with locking device
  • Covered rack keeps gear teeth clean
  • Thumb roller included
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty
Catalog No. 50-005-28160 Supplier MSC Supplier No. 37530631
The dial caliper is a precision measuring instrument designed for accurate inside, outside, depth, and step measurements. Constructed from stainless steel, it offers an 18-inch maximum measurement range with a 0.001 inch graduation and 0.002 inch accuracy. This caliper features preloaded dual pinions for consistent repeatability and comes with NIST traceability certification.
  • Stainless steel construction with surface-hardened measuring surfaces
  • Preloaded dual pinions eliminate backlash and provide better repeatability
  • 4-Way measurements: inside, outside, depth and step
  • Easy zero adjustment per revolution
  • Adjustable bezel with locking device
  • Covered rack keeps gear teeth clean
  • Thumb roller included
  • Short form calibration certificate included
  • Furnished in fitted case
  • Two year limited warranty