Learn More
MSC SPI Dial Caliper, 6 in, 0.001 in Graduation, Stainless Steel Jaws
Catalog No. 5000528353
Encompass
This precision measuring tool offers a 6 in maximum measurement capacity with 0.001 in graduation and 0.001 in accuracy. It performs outside, step, inside, and depth measurements. The dial provides 0.100 in per revolution for accurate readings. Constructed with durable stainless steel, it ensures longevity and reliable performance. This caliper is calibrated and includes NIST traceability certification. It comes with a fitted case, and its blue dial face enhances readability.
  • Precision measuring tool for outside, step, inside, and depth measurements.
  • 6 in maximum measurement capacity.
  • 0.001 in graduation.
  • 0.100 in per revolution.
  • Accuracy of 0.001 in.
  • Durable stainless steel construction.
  • Inside and outside jaw styles.
  • NIST traceability certified.
  • Comes with a fitted case.
  • Blue dial face for enhanced readability.
Catalog No. 50-005-28353 Supplier MSC Supplier No. 37530557
Add to Cart
Add to Cart
This precision measuring tool offers a 6 in maximum measurement capacity with 0.001 in graduation and 0.001 in accuracy. It performs outside, step, inside, and depth measurements. The dial provides 0.100 in per revolution for accurate readings. Constructed with durable stainless steel, it ensures longevity and reliable performance. This caliper is calibrated and includes NIST traceability certification. It comes with a fitted case, and its blue dial face enhances readability.
  • Precision measuring tool for outside, step, inside, and depth measurements.
  • 6 in maximum measurement capacity.
  • 0.001 in graduation.
  • 0.100 in per revolution.
  • Accuracy of 0.001 in.
  • Durable stainless steel construction.
  • Inside and outside jaw styles.
  • NIST traceability certified.
  • Comes with a fitted case.
  • Blue dial face for enhanced readability.