Learn More
MSC SPI Dial Caliper, 6 inch, 0.001 inch Graduation, Stainless Steel
Catalog No. 5000527939
Encompass
This stainless steel dial caliper features surface-hardened measuring surfaces for durability and precision. It offers versatile 4-way measurements (inside, outside, depth, step) and incorporates preloaded dual pinions to eliminate backlash and enhance repeatability. Key features include an easy zero adjustment, an adjustable bezel with locking device, and a covered rack to protect gear teeth. It is supplied with a thumb roller, a short-form Calibration Certificate, and comes in a fitted case.
  • Preloaded dual pinions for enhanced repeatability
  • Easy zero adjustment
  • Adjustable bezel with locking device
  • Covered rack keeps gear teeth clean
  • Thumb roller included
  • Short-form Calibration Certificate
  • Fitted case for storage
  • Two-year limited warranty
Catalog No. 50-005-27939 Supplier MSC Supplier No. 37530722
This stainless steel dial caliper features surface-hardened measuring surfaces for durability and precision. It offers versatile 4-way measurements (inside, outside, depth, step) and incorporates preloaded dual pinions to eliminate backlash and enhance repeatability. Key features include an easy zero adjustment, an adjustable bezel with locking device, and a covered rack to protect gear teeth. It is supplied with a thumb roller, a short-form Calibration Certificate, and comes in a fitted case.
  • Preloaded dual pinions for enhanced repeatability
  • Easy zero adjustment
  • Adjustable bezel with locking device
  • Covered rack keeps gear teeth clean
  • Thumb roller included
  • Short-form Calibration Certificate
  • Fitted case for storage
  • Two-year limited warranty