Learn More
MSC SPI Dial Depth Gage, 0 to 4 in, 0.001 in, Steel
Catalog No. 5000533672
Encompass
This dial depth gage provides precise measurements from 0 to 4 inches with a 0.001 inch graduation and 0.002 inch accuracy. Constructed from durable steel, it features a 4 inch base measuring length and a clear white dial face, making it suitable for a variety of industrial and laboratory applications.
  • Lifting lever for easy operation
  • Hardened base for durability
  • Revolution counter for accurate readings
  • Precision ground for enhanced accuracy
  • Includes a fitted case for protection and storage
Catalog No. 50-005-33672 Supplier MSC Supplier No. 37530862
This dial depth gage provides precise measurements from 0 to 4 inches with a 0.001 inch graduation and 0.002 inch accuracy. Constructed from durable steel, it features a 4 inch base measuring length and a clear white dial face, making it suitable for a variety of industrial and laboratory applications.
  • Lifting lever for easy operation
  • Hardened base for durability
  • Revolution counter for accurate readings
  • Precision ground for enhanced accuracy
  • Includes a fitted case for protection and storage