Learn More
MSC SPI Dial Drop Indicator, 0.001 in Graduation, 0-100 Dial Reading, 2-1/4 in Diameter, White
Catalog No. 5000533187
Encompass
This dial drop indicator is a precision measuring instrument designed for various industrial and laboratory applications. It provides accurate readings with specific graduation and dial range capabilities.
  • 0.001 in graduation
  • Lug back type
  • 0-100 dial reading
  • 2-1/4 in dial diameter
  • White dial color
  • Made in Germany
Catalog No. 50-005-33187 Supplier MSC Supplier No. 08368946
This dial drop indicator is a precision measuring instrument designed for various industrial and laboratory applications. It provides accurate readings with specific graduation and dial range capabilities.
  • 0.001 in graduation
  • Lug back type
  • 0-100 dial reading
  • 2-1/4 in dial diameter
  • White dial color
  • Made in Germany