Learn More
MSC SPI Dial Drop Indicator, 0.0500 in Max Measurement, 0-10 Dial Reading, 0.0003 in Accuracy
Catalog No. 5000534076
Encompass
This dial drop indicator is designed to measure the geometric tolerances and dimensional deviations of workpieces.
  • Measures geometric tolerances and dimensional deviations of workpieces.
  • Offers an accuracy of 0.0003 "
  • Maximum measurement capacity is 0.0500 "
  • Features a 0-10 dial reading.
  • Equipped with a revolution counter.
  • Includes a counter pointer for easy reading.
  • Provides universal positioning capability.
  • Supplied with NIST traceability certification.
  • Has a dial graduation of 0.000100 "
  • Designed with lug and flat back types.
  • Features jeweled bearing type.
Catalog No. 50-005-34076 Supplier MSC Supplier No. 38000063
This dial drop indicator is designed to measure the geometric tolerances and dimensional deviations of workpieces.
  • Measures geometric tolerances and dimensional deviations of workpieces.
  • Offers an accuracy of 0.0003 "
  • Maximum measurement capacity is 0.0500 "
  • Features a 0-10 dial reading.
  • Equipped with a revolution counter.
  • Includes a counter pointer for easy reading.
  • Provides universal positioning capability.
  • Supplied with NIST traceability certification.
  • Has a dial graduation of 0.000100 "
  • Designed with lug and flat back types.
  • Features jeweled bearing type.