Learn More
MSC SPI Dial Drop Indicator, 0-100 Dial Reading, 0.001 in Graduation, 2-1/4 in Dial Diameter, Lug Back
Catalog No. 5000529098
Encompass
The SPI Dial Drop Indicator is engineered for precise measurement applications, featuring a clear white dial with a 0-100 reading and a fine 0.001 inch graduation. Designed with a durable lug back and a 2-1/4 inch dial diameter, it offers an overall travel of two inches, making it suitable for a variety of tasks requiring accurate displacement readings.
  • Lug back for secure mounting
  • White dial for clear visibility
  • 0-100 dial reading for versatile measurements
  • 2-1/4 inch dial diameter
  • 0.001 inch graduation for high precision
  • Two inches of overall travel
Catalog No. 50-005-29098 Supplier MSC Supplier No. 08368938
The SPI Dial Drop Indicator is engineered for precise measurement applications, featuring a clear white dial with a 0-100 reading and a fine 0.001 inch graduation. Designed with a durable lug back and a 2-1/4 inch dial diameter, it offers an overall travel of two inches, making it suitable for a variety of tasks requiring accurate displacement readings.
  • Lug back for secure mounting
  • White dial for clear visibility
  • 0-100 dial reading for versatile measurements
  • 2-1/4 inch dial diameter
  • 0.001 inch graduation for high precision
  • Two inches of overall travel