Learn More
MSC SPI Dial Drop Indicator, 0.1000 in Max Measurement, 0-100 Dial Reading, Jeweled Bearing
Catalog No. 5000527528
Encompass
This dial drop indicator offers precise measurements with a maximum range of 0.1000 inches and a 0-100 dial reading. It features a white dial, a graduation of 0.001000 inches, and a plunger back with a 4-48 thread contact point.
  • Fast, accurate set-ups
  • Lever attachment
  • Universal clamp included
  • #4-48 thread size on contact point
  • Fully jeweled test indicator
  • 5/16 in x 5 in holding rod
  • G clamp
  • Tool post holder
  • 5/16 in x 3 in holding rod
Catalog No. 50-005-27528 Supplier MSC Supplier No. 76422369
This dial drop indicator offers precise measurements with a maximum range of 0.1000 inches and a 0-100 dial reading. It features a white dial, a graduation of 0.001000 inches, and a plunger back with a 4-48 thread contact point.
  • Fast, accurate set-ups
  • Lever attachment
  • Universal clamp included
  • #4-48 thread size on contact point
  • Fully jeweled test indicator
  • 5/16 in x 5 in holding rod
  • G clamp
  • Tool post holder
  • 5/16 in x 3 in holding rod