Learn More
MSC SPI Dial Drop Indicator, 0.2000 in Max Measurement, 0.0001 in Graduation, Revolution Counter
Catalog No. 5000536257
Encompass
This dial drop indicator offers precise measurements for various applications. It features a flat back for easy mounting and a revolution counter for accurate readings over its full range. Designed for reliability, it includes traceable calibration and robust housing.
  • Maximum measurement of 0.2000 in.
  • Dial graduation of 0.0001 in.
  • Dial reading options of 0-50-0 and 0-100.
  • Flat back type for stable positioning.
  • Revolution counter for tracking turns.
  • Rigid, forged brass housing for durability.
  • Includes tolerance pointers and both flat and lug back.
  • Certificate of calibration traceable to NPL.
  • Complies with ASME B89.1.10M-2001 and AGD Group 2 standards.
Catalog No. 50-005-36257 Supplier MSC Supplier No. 06543870
This dial drop indicator offers precise measurements for various applications. It features a flat back for easy mounting and a revolution counter for accurate readings over its full range. Designed for reliability, it includes traceable calibration and robust housing.
  • Maximum measurement of 0.2000 in.
  • Dial graduation of 0.0001 in.
  • Dial reading options of 0-50-0 and 0-100.
  • Flat back type for stable positioning.
  • Revolution counter for tracking turns.
  • Rigid, forged brass housing for durability.
  • Includes tolerance pointers and both flat and lug back.
  • Certificate of calibration traceable to NPL.
  • Complies with ASME B89.1.10M-2001 and AGD Group 2 standards.