Learn More
MSC SPI Dial Height Gage, 20 in, Stainless Steel, 0.001 in Graduation, Dial & Counter
Catalog No. 5000531001
Encompass
The 20 in Stainless Steel Dial Height Gage features 0.001 in graduation and is accurate to 0.002 in. It includes a dial and counter display, and comes with NIST Traceability Certification. The dial and counter design facilitates accurate and easy-to-read fine and coarse measurements, with a resettable digital counter that has two displays for individual upward and downward measurements from a preset zero.
  • Accurate to 0.002 in
  • Dial and counter display
  • NIST traceability certification
  • Allows for accurate and easy-to-read fine and coarse measurements
  • Resettable digital counter has two displays for individual upward and downward measurements from a preset zero
Catalog No. 50-005-31001 Supplier MSC Supplier No. 69549160
The 20 in Stainless Steel Dial Height Gage features 0.001 in graduation and is accurate to 0.002 in. It includes a dial and counter display, and comes with NIST Traceability Certification. The dial and counter design facilitates accurate and easy-to-read fine and coarse measurements, with a resettable digital counter that has two displays for individual upward and downward measurements from a preset zero.
  • Accurate to 0.002 in
  • Dial and counter display
  • NIST traceability certification
  • Allows for accurate and easy-to-read fine and coarse measurements
  • Resettable digital counter has two displays for individual upward and downward measurements from a preset zero