Learn More
MSC SPI Dial Indicator Contact Point, Chrome Alloy Steel, #4-48 Thread
Catalog No. 5000511658
Encompass
These contact points are designed for use with dial indicators. They feature a ball needle style made from chrome alloy steel, offering precision for specific measurement tasks. These points are ideal for indicator applications and are not intended for sizing or gauging where side pressure may occur.
  • Designed for use with dial indicators.
  • Features a ball needle contact point style.
  • Constructed from chrome alloy steel.
  • Has a #4-48 thread size.
  • Provides a 0.7500 in point length.
  • Equipped with a 0.0469 in ball diameter.
  • Meets AGD Group 1 industry designation.
  • Not recommended for sizing or gauging applications where side pressure would be present.
Catalog No. 50-005-11658 Supplier MSC Supplier No. 35915271
These contact points are designed for use with dial indicators. They feature a ball needle style made from chrome alloy steel, offering precision for specific measurement tasks. These points are ideal for indicator applications and are not intended for sizing or gauging where side pressure may occur.
  • Designed for use with dial indicators.
  • Features a ball needle contact point style.
  • Constructed from chrome alloy steel.
  • Has a #4-48 thread size.
  • Provides a 0.7500 in point length.
  • Equipped with a 0.0469 in ball diameter.
  • Meets AGD Group 1 industry designation.
  • Not recommended for sizing or gauging applications where side pressure would be present.