Learn More
MSC SPI Dial Indicator Kit, 1 in Range, 0.001 in Graduations, White Dial
Catalog No. 5000533365
Encompass
This kit combines a deluxe dial indicator with a robust magnetic base, providing a comprehensive solution for precise measurement tasks. The indicator features a 1 inch range and 0.001 inch graduations, ideal for demanding applications. The magnetic base offers strong holding power and versatile mounting options, ensuring stability and adaptability in various work environments. All components are supplied in a protective fitted case for convenience and durability.
  • Accuracy meets or exceeds Federal Specifications.
  • Easy reading graduations and adjustable bezel for zero setting.
  • Calibration certificate traceable to NIST included.
  • Magnetic base offers 45 lbs. of holding power.
  • Zinc diecast housing for the magnetic base.
  • Multiple mounting options on the magnetic base.
  • Furnished in a fitted case for protection and portability.
Catalog No. 50-005-33365 Supplier MSC Supplier No. 02830107
This kit combines a deluxe dial indicator with a robust magnetic base, providing a comprehensive solution for precise measurement tasks. The indicator features a 1 inch range and 0.001 inch graduations, ideal for demanding applications. The magnetic base offers strong holding power and versatile mounting options, ensuring stability and adaptability in various work environments. All components are supplied in a protective fitted case for convenience and durability.
  • Accuracy meets or exceeds Federal Specifications.
  • Easy reading graduations and adjustable bezel for zero setting.
  • Calibration certificate traceable to NIST included.
  • Magnetic base offers 45 lbs. of holding power.
  • Zinc diecast housing for the magnetic base.
  • Multiple mounting options on the magnetic base.
  • Furnished in a fitted case for protection and portability.