Learn More
MSC SPI Dial Test Indicator & Base Kit, 0.030 in Max Measurement, 0.0005 in Graduation, White Dial
Catalog No. 5000529818
Encompass
This Dial Test Indicator and Base Kit provides precise measurements. It features a maximum measurement of 0.030 inches, a 0-15-0 dial reading, a white dial color, and a graduation of 0.0005 inches.
  • Fits milling machines.
  • Kit includes holder and indicator.
Catalog No. 50-005-29818 Supplier MSC Supplier No. 01579382
This Dial Test Indicator and Base Kit provides precise measurements. It features a maximum measurement of 0.030 inches, a 0-15-0 dial reading, a white dial color, and a graduation of 0.0005 inches.
  • Fits milling machines.
  • Kit includes holder and indicator.