Learn More
MSC SPI Dial Test Indicator & Base Kit, 0.0300 in Max Measurement, 0-15-0 Dial
Catalog No. 5000513813
Encompass
This dial test indicator and base kit is designed for precise measurements in various applications. It features a maximum measurement capacity of 0.0300 inches and a fine dial graduation of 0.000500 inches, along with a 0-15-0 dial reading, providing clear and accurate readings. The kit includes a magnetic base for stable and versatile positioning during use.
  • Indicator type is dial test
  • Magnetic base for stable positioning
  • Maximum measurement of 0.0300 inches
  • Dial graduation of 0.000500 inches
  • 0-15-0 dial reading for easy interpretation
  • Equipped with an on/off switch for convenience
Catalog No. 50-005-13813 Supplier MSC Supplier No. 06422976
This dial test indicator and base kit is designed for precise measurements in various applications. It features a maximum measurement capacity of 0.0300 inches and a fine dial graduation of 0.000500 inches, along with a 0-15-0 dial reading, providing clear and accurate readings. The kit includes a magnetic base for stable and versatile positioning during use.
  • Indicator type is dial test
  • Magnetic base for stable positioning
  • Maximum measurement of 0.0300 inches
  • Dial graduation of 0.000500 inches
  • 0-15-0 dial reading for easy interpretation
  • Equipped with an on/off switch for convenience