Learn More
MSC SPI Digital Chamfer Gage, 0 to 1 in, 0 to 90°, 0.0001 in
Catalog No. 5000510891
Encompass
The SPI Digital Chamfer Gage is designed for precise measurements, featuring a digital display and a 3-blade plunger for accurate readings. Constructed from hardened tool steel, this gage offers a measurement range of 0 to 1 inch and an included angle range of 0 to 90 degrees, with an accuracy of 0.0001 "
  • Digital display for easy and accurate readings.
  • 3-blade plunger ensures precise measurement.
  • Hardened tool steel construction for durability.
  • Measurement range of 0 to 1 "
  • Included angle range of 0 to 90 degrees.
  • High precision with 0.0001 inch accuracy.
Catalog No. 50-005-10891 Supplier MSC Supplier No. 79074068
The SPI Digital Chamfer Gage is designed for precise measurements, featuring a digital display and a 3-blade plunger for accurate readings. Constructed from hardened tool steel, this gage offers a measurement range of 0 to 1 inch and an included angle range of 0 to 90 degrees, with an accuracy of 0.0001 "
  • Digital display for easy and accurate readings.
  • 3-blade plunger ensures precise measurement.
  • Hardened tool steel construction for durability.
  • Measurement range of 0 to 1 "
  • Included angle range of 0 to 90 degrees.
  • High precision with 0.0001 inch accuracy.