Learn More
MSC SPI Digital Chamfer Gage, 1 to 2 in, 0 to 90°, 0.0001 in
Catalog No. 5000528163
Encompass
The SPI Digital Chamfer Gage is designed for accurate measurement of chamfers. It features a 3-blade plunger, digital display, and provides precise measurements with 0.0001 inch accuracy for chamfers ranging from 1 to 2 inches and angles from 0 to 90 degrees.
  • 0.0001 inch accuracy
  • 3-blade plunger
  • Digital display
  • SPC output port
Catalog No. 50-005-28163 Supplier MSC Supplier No. 37936002
The SPI Digital Chamfer Gage is designed for accurate measurement of chamfers. It features a 3-blade plunger, digital display, and provides precise measurements with 0.0001 inch accuracy for chamfers ranging from 1 to 2 inches and angles from 0 to 90 degrees.
  • 0.0001 inch accuracy
  • 3-blade plunger
  • Digital display
  • SPC output port