Learn More
MSC SPI Digital Electronic Drop Indicator, 1 in, 0.000150 in Accuracy, IP65
Catalog No. 5000533774
Encompass
This SPI Digital Electronic Drop Indicator offers precise measurements for various applications. It features a 1-in maximum range with an accuracy of 0.000150 in, ensuring high-resolution readings. Designed with an IP65 protection level, it is resistant to dust and water, making it suitable for demanding environments.
  • MM/inch measurement capability.
  • Zero setting function for relative measurements.
  • Preset function for convenience.
  • ABS/INC mode for absolute and incremental measurements.
  • Tolerance setting for quality control.
  • Data output supported for external connectivity.
  • Protection level IP65 for dust and water resistance.
  • Calibrated for accuracy assurance.
  • Intuitive reading for ease of use.
  • High resolution for precise measurements.
  • Easy operation for user-friendliness.
  • Includes NIST traceability certification.
Catalog No. 50-005-33774 Supplier MSC Supplier No. 37998028
This SPI Digital Electronic Drop Indicator offers precise measurements for various applications. It features a 1-in maximum range with an accuracy of 0.000150 in, ensuring high-resolution readings. Designed with an IP65 protection level, it is resistant to dust and water, making it suitable for demanding environments.
  • MM/inch measurement capability.
  • Zero setting function for relative measurements.
  • Preset function for convenience.
  • ABS/INC mode for absolute and incremental measurements.
  • Tolerance setting for quality control.
  • Data output supported for external connectivity.
  • Protection level IP65 for dust and water resistance.
  • Calibrated for accuracy assurance.
  • Intuitive reading for ease of use.
  • High resolution for precise measurements.
  • Easy operation for user-friendliness.
  • Includes NIST traceability certification.