Learn More
MSC SPI Digital Force Gage, 100 N Capacity, 0.01 N Resolution, ±0.2% F.S. Accuracy
Catalog No. 5000535489
Encompass
SPI Digital Force Gages are designed for precise measurement applications. The ASFG 100 model offers a 100 N capacity and a fine resolution of 0.01 N. With an accuracy of ±0.2% F.S., it provides reliable results for various force testing needs.
  • 100 N load capacity
  • 0.01 N resolution
  • ±0.2% F.S. accuracy
  • Measurement modes: track, peak, preset peak
Catalog No. 50-005-35489 Supplier MSC Supplier No. 85388155
SPI Digital Force Gages are designed for precise measurement applications. The ASFG 100 model offers a 100 N capacity and a fine resolution of 0.01 N. With an accuracy of ±0.2% F.S., it provides reliable results for various force testing needs.
  • 100 N load capacity
  • 0.01 N resolution
  • ±0.2% F.S. accuracy
  • Measurement modes: track, peak, preset peak